1-(2-Hydroxyethyl)pyrrole 99%

Supplier: APOLLO SCIENTIFIC

Warning

OR200029-250MG
76569-924EA 30 USD
76569-924
1-(2-Hydroxyethyl)pyrrole 99%
1-(2-Hydroxyethyl)pyrrole
Formula: C₆H₉NO
Boiling Pt: 215 °C
Density: 1.06 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00191451
CAS Number: 6719-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR