3-Bromo-7-methylimidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC

Warning

OR1978-1G OR1978-5G
76569-890EA 224 USD
76569-890 76569-888
3-Bromo-7-methylimidazo[1,2-a]pyridine
3-Bromo-7-methylimidazo[1,2-a]pyridine
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056208
CAS Number: 56051-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR