2,4-Dimethylbenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR19563-500G OR19563-100G OR19563-25G
76569-860EA 139 USD
76569-860 76569-858 76569-856
2,4-Dimethylbenzoic acid
2,4-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 267 °C
Melting Pt: 124…126 °C
Flash Pt: 154 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002480
CAS Number: 611-01-8
EINECS: 210-246-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR