Iminodiacetonitrile, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

OR18385-2.5KG OR18385-500G
76569-450EA 352 USD
76569-450 76569-448
Iminodiacetonitrile, Technical Grade
Iminodiacetonitrile
Formula: C₄H₅N₃
MW: 95.11 g/mol
Melting Pt: 69…71 °C
Storage Temperature: Ambient
EINECS: 211-058-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR