2,5-Dimercapto-1,3,4-thiadiazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3,4-Thiadiazole-2,5-dithiol

OR18357-250G
76569-388EA 28 USD
76569-388
2,5-Dimercapto-1,3,4-thiadiazole
2,5-Dimercapto-1,3,4-thiadiazole
Formula: C₂H₂N₂S₃
MW: 150.25 g/mol
Melting Pt: 161…164 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00003103
CAS Number: 1072-71-5
EINECS: 214-014-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR