4-tert-Butyl-1-bromobenzene 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-Bromo-4-(tert-butyl)benzene2-(4-Bromophenyl)-2-methylpropane4-Bromo-tert-butylbenzene1-Bromo-4-tert-butylbenzene

OR18318-500G OR18318-1KG OR18318-100G
76569-196EA 21 USD
76569-196 76569-200 76569-198
4-tert-Butyl-1-bromobenzene 97%
4-tert-Butyl-1-bromobenzene
Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 106…109 °C/20mmHg
Melting Pt: 15…16 °C
Density: 1.229 g/cm³ (25 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00000108
CAS Number: 3972-65-4
EINECS: 223-599-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR