1-(Hydroxymethyl)-5,5-dimethylhydantoin 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 5,5-Dimethyl-1-(hydroxymethyl)hydantoin

OR18297-500G OR18297-25G OR18297-100G
76569-130EA 13 USD
76569-130 76569-134 76569-132
1-(Hydroxymethyl)-5,5-dimethylhydantoin 97%
1-(Hydroxymethyl)-5,5-dimethylhydantoin
Formula: C₆H₁₀N₂O₃
MW: 158.16
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00020866
CAS Number: 116-25-6
EINECS: 204-132-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR