2-Quinolinol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-QuinoloneQuinolin-2(1H)-one2-HydroxyquinolineCarbostyrilQuinolin-2-ol

OR18280-5G OR18280-2.5G OR18280-25G
76569-098EA 58 USD
76569-098 76569-096 76569-094
2-Quinolinol 98%
2-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 198…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00006743
CAS Number: 59-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR