1-Methylhydantoin 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR18274-1G
76569-076EA 13 USD
76569-076
1-Methylhydantoin 97%
1-Methylhydantoin
Formula: C₄H₆N₂O₂
Melting Pt: 156…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00003187
CAS Number: 616-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR