4-Bromo-1-iodo-2-nitrobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromo-2-iodonitrobenzene

OR17993-100G OR17993-5G OR17993-25G
76568-960EA 42 USD
76568-960 76568-958 76568-962
4-Bromo-1-iodo-2-nitrobenzene 99%
4-Bromo-1-iodo-2-nitrobenzene
Formula: C₆H₃BrINO₂
MW: 327.9020 g/mol
Melting Pt: 87…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11505492
CAS Number: 112671-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR