Ethyl-4-amino-2-chlorobenzoate 95+%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Ethyl 4-amino-2-chlorobenzoate
OR17937-1G
OR17937-500MG
76568-956EA
19
USD
76568-956
76568-954
Ethyl-4-amino-2-chlorobenzoate 95+%
Ethyl 4-amino-2-chlorobenzoate
Formula:
C₉H₁₀ClNO₂ MW: 199.6342 g/mol Melting Pt: 109…111 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09034640 CAS Number: 16017-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...