Methyl-5-methyl-2-nitrobenzoate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 6-Nitro-m-toluic acid methyll esterMethyl 5-methyl-2-nitrobenzoate5-Methyl-2-nitrobenzoic acid methyll esterMethyl 6-Nitro-m-toluate

OR17822-250MG OR17822-1G
76568-840EA 49 USD
76568-840 76568-842
Methyl-5-methyl-2-nitrobenzoate
Methyl 5-methyl-2-nitrobenzoate
Formula: C₉H₉NO₄
MW: 195.1721 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00157275
CAS Number: 20587-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR