6-Bromo-2-chloroquinoline

Supplier: APOLLO SCIENTIFIC

Warning

OR17558-1G OR17558-5G
76568-690EA 19 USD
76568-690 76568-692
6-Bromo-2-chloroquinoline
6-Bromo-2-chloroquinoline
Formula: C₉H₅BrClN
MW: 242.50 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD04115272
CAS Number: 1810-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR