2-Amino-6-bromobenzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR17502-25G OR17502-100G OR17502-5G
76568-652EA 22 USD
76568-652 76568-654 76568-656
2-Amino-6-bromobenzonitrile 97%
2-Amino-6-bromobenzonitrile
Formula: C₇H₅BrN₂
MW: 197.02 g/mol
Melting Pt: 152…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834769
CAS Number: 77326-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR