2-Chloroquinazoline

Supplier: APOLLO SCIENTIFIC

Warning

OR17301-5G OR17301-1G OR17301-250MG
76568-604EA 70 USD
76568-604 76568-606 76568-602
2-Chloroquinazoline
2-Chloroquinazoline
Formula: C₈H₅ClN₂
MW: 164.5900 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09959715
CAS Number: 6141-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR