2-Bromo-5-iodonitrobenzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-4-Iodo-2-nitrobenzene

OR17118-1G OR17118-250MG OR17118-5G
76568-560EA 79 USD
76568-560 76568-556 76568-558
2-Bromo-5-iodonitrobenzene 97%
2-Bromo-5-iodonitrobenzene
Formula: C₆H₃BrINO₂
MW: 327.9000 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD12547765
CAS Number: 713512-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR