3-(3-Chlorophenyl)-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Danger

OR17-250MG OR17-1G
76568-486EA 45 USD
76568-486 76568-488
3-(3-Chlorophenyl)-1H-pyrazole
3-(3-Chlorophenyl)-1H-pyrazole
Formula: C₉H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD09860599
CAS Number: 59843-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR