3,4,6-Trichloropyridazine

Supplier: APOLLO SCIENTIFIC

Warning

OR16890-25G OR16890-5G OR16890-100G
76568-386EA 22 USD
76568-386 76568-388 76568-384
3,4,6-Trichloropyridazine
3,4,6-Trichloropyridazine
Formula: C₄HCl₃N₂
MW: 183.4231 g/mol
Boiling Pt: 96…97.5 °C/2mmHg
Melting Pt: 55…59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00834957
CAS Number: 6082-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR