3-Iodo-4-nitro-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR16867-500MG OR16867-1G
76568-364EA 58 USD
76568-364 76568-362
3-Iodo-4-nitro-1H-indazole
3-Iodo-4-nitro-1H-indazole
Formula: C₇H₄IN₃O₂
MW: 289.0300 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781574
CAS Number: 885521-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR