3-Bromo-5-chloropyridine-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromo-5-chloropicolinic acid

OR16596-5G OR16596-1G
76568-230EA 104 USD
76568-230 76568-228
3-Bromo-5-chloropyridine-2-carboxylic acid
3-Bromo-5-chloropyridine-2-carboxylic acid
Formula: C₆H₃BrClNO₂
MW: 236.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827551
CAS Number: 1189513-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR