(±)-Tetrahydrofuran-3-carboxylic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Tetrahydro-3-furoic acid
, Tetrahydrofuran-3-carboxylic acid
OR16452-1G
OR16452-25G
OR16452-5G
76568-174EA
36
USD
76568-174
76568-176
76568-172
(±)-Tetrahydrofuran-3-carboxylic acid
(±)-Tetrahydro-2-furoic acid
Formula:
C₅H₈O₃ MW: 116.1152 g/mol Boiling Pt: 140…141 °C/15mmHg Density: 1.214 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075173 CAS Number: 89364-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...