2-Chloro-4-hydroxybenzonitrile 98+%

Supplier: APOLLO SCIENTIFIC
Warning

OR16169-5G OR16169-1G OR16169-25G
76568-122EA 159 USD
76568-122 76568-120 76568-118
2-Chloro-4-hydroxybenzonitrile 98+%
2-Chloro-4-hydroxybenzonitrile
Formula: C₇H₄ClNO
MW: 153.57 g/mol
Melting Pt: 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00052185
CAS Number: 3336-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR