(S)-(+)-1-Boc-3-hydroxypyrrolidine 99%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate(S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

OR15680-10G OR15680-100G OR15680-25G
76567-906EA 80 USD
76567-906 76567-904 76567-902
(S)-(+)-1-Boc-3-hydroxypyrrolidine 99%
(S)-(+)-1-Boc-3-hydroxypyrrolidine

Chiral intermediate for pharmaceuticals and agrochemicals.

Formula: C₉H₁₇NO₃
MW: 187.2362 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01317839
CAS Number: 101469-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR