Methyl trans-4-bromocinnamate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl trans-4-BromocinnamateMethyl (2E)-3-(4-bromophenyl)prop-2-enoateMethyl (E)-3-(4-bromophenyl)acrylate

OR15501-5G OR15501-1G OR15501-250MG
76567-780EA 82 USD
76567-780 76567-778 76567-776
Methyl trans-4-bromocinnamate 98%
Methyl trans-4-bromocinnamate
Formula: C₁₀H₉BrO₂
MW: 241.09 g/mol
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00157162
CAS Number: 71205-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR