2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4',5'-Dihydro-1H,3'H-2,2'-bipyrrole

OR15487-1G OR15487-250MG
76567-774EA 220 USD
76567-774 76567-772
2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole
2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole
Formula: C₈H₁₀N₂
MW: 134.18 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00030437
CAS Number: 10087-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR