2-Phenyl-1,3-thiazole-4-carbohydrazide

Supplier: APOLLO SCIENTIFIC

Warning

OR15428-1G OR15428-5G
76567-738EA 519 USD
76567-738 76567-736
2-Phenyl-1,3-thiazole-4-carbohydrazide
2-Phenyl-1,3-thiazole-4-carbohydrazide
Formula: C₁₀H₉N₃OS
Melting Pt: 136…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00174011
CAS Number: 7113-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR