Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC

Warning

OR15417-1G
76567-734EA 69 USD
76567-734
Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate
Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate
Formula: C₁₀H₁₁N₅O₂
MW: 233.23 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00838109
CAS Number: 23947-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR