4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Thiomorpholin-4-yl)benzaldehyde

OR15298-1G OR15298-5G
76567-672EA 643 USD
76567-672 76567-670
4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde
4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde
Formula: C₁₁H₁₃NOS
MW: 207.3 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD08276967
CAS Number: 27913-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR