Ethyl-2-aminonicotinate 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Ethyl 2-aminonicotinate

OR15285-5G OR15285-25G
76567-650EA 61 USD
76567-650 76567-648
Ethyl-2-aminonicotinate 98%
Ethyl 2-aminonicotinate
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 266.75 °C
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01569452
CAS Number: 13362-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR