1-(5-Chloro-2-nitrophenyl)-1-ethanone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5'-Chloro-2'-nitroacetophenone1-(5-Chloro-2-nitrophenyl)ethanone

OR15281-1G
76567-646EA 13 USD
76567-646
1-(5-Chloro-2-nitrophenyl)-1-ethanone
1-(5-Chloro-2-nitrophenyl)-1-ethanone
Formula: C₈H₆ClNO₃
MW: 199.59 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD03787322
CAS Number: 18640-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR