2-(Chloromethyl)-1-methyl-1H-imidazole
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR15254-1G
OR15254-500MG
OR15254-100MG
76567-610EA
76
USD
76567-610
76567-614
76567-612
2-(Chloromethyl)-1-methyl-1H-imidazole
2-(Chloromethyl)-1-methyl-1H-imidazole
Formula:
C₅H₇ClN₂ Melting Pt: 182…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234119 CAS Number: 19225-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...