2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR15224-1G
76567-578EA 41 USD
76567-578
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Formula: C₁₀H₉NO₄
MW: 207.19 g/mol
Melting Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD04124544
CAS Number: 154365-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR