Methyl-4-amino-3-bromo-5-nitrobenzoate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Methyl 4-amino-3-bromo-5-nitrobenzoate
OR15218-1G
OR15218-25G
OR15218-5G
76567-576EA
288
USD
76567-576
76567-574
76567-572
Methyl-4-amino-3-bromo-5-nitrobenzoate
Methyl 4-amino-3-bromo-5-nitrobenzoate
Formula:
C₈H₇BrN₂O₄ MW: 275.06 g/mol Melting Pt: 108…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD08443938 CAS Number: 105655-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...