Potassium-4-iodobenzenesulfonate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Potassium 4-iodobenzenesulfonatePotassium 4-iodobenzenesulphonate

OR15122-5G OR15122-1G
76567-484EA 322 USD
76567-484 76567-482
Potassium-4-iodobenzenesulfonate
Potassium-4-iodobenzenesulfonate
Formula: C₆H₄IKO₃S
MW: 322.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656232
CAS Number: 13035-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR