tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-[(Dimethylamino)methylene]piperidin-4-one, N1-BOC protected

OR15082-10G OR15082-1G OR15082-5G
76567-454EA 189 USD
76567-454 76567-452 76567-450
tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-3-[(dimethylamino)methylene]-4-oxotetrahydro-1(2H)-pyridinecarboxylate
Formula: C₁₃H₂₂N₂O₃
MW: 254.33 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD08443986
CAS Number: 157327-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR