5-Bromo-7-nitro-1H-indazole

Supplier: APOLLO SCIENTIFIC

Warning

OR15069-1G
76567-444EA 63 USD
76567-444
5-Bromo-7-nitro-1H-indazole
5-Bromo-7-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD03617604
CAS Number: 316810-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR