2-Mercapto-1,3,4-thiadiazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1,3,4-Thiadiazole-2-thiol

OR15040-5G
76560-636EA 13 USD
76560-636
2-Mercapto-1,3,4-thiadiazole
2-Mercapto-1,3,4-thiadiazole
Formula: C₂H₂N₂S₂
MW: 118.17 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00040404
CAS Number: 18686-82-3
EINECS: 242-504-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR