(+)-Di-p-toluoyl-D-tartaric acid 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid
, Di-p-toluoyl-D-tartaric Acid
, Di-p-toluoyl-D-tartaric acid
OR14900-100G
OR14900-500G
OR14900-250G
76567-278EA
26
USD
76567-278
76567-276
76567-280
(+)-Di-p-toluoyl-D-tartaric acid 99%
(+)-Di-p-toluoyl-D-tartaric acid
Optical resolving agent.
Formula:
C₂₀H₁₈O₈ MW: 386.36 g/mol Melting Pt: 169…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008552 CAS Number: 32634-68-7 EINECS: 251-132-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...