Methyl-5-bromosalicylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 5-bromo-2-hydroxybenzoate

OR14417-100G OR14417-25G OR14417-5G
76567-056EA 164 USD
76567-056 76567-054 76567-052
Methyl-5-bromosalicylate
Methyl 5-bromosalicylate
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00016460
CAS Number: 4068-76-2
EINECS: 223-776-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR