N-(3-Methoxyphenyl)urea

Supplier: APOLLO SCIENTIFIC
Warning

OR14387-1G OR14387-500MG OR14387-250MG
76567-030EA 152 USD
76567-030 77404-938 76567-028
N-(3-Methoxyphenyl)urea
N-(3-Methoxyphenyl)urea
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 138…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00025423
CAS Number: 139-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR