Methyl-6-amino-5-bromonicotinate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 6-amino-5-bromonicotinate

OR14385-5G OR14385-1G
76567-026EA 86 USD
76567-026 76567-024
Methyl-6-amino-5-bromonicotinate
Methyl 6-amino-5-bromonicotinate
Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD06659561
CAS Number: 180340-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR