4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,2-Dihydro-2,5-dimethyl-4-(2-hydroxyethyl)-3H-pyrazol-3-one

OR14377-250MG
76567-022EA 57.04 USD
76567-022
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one
Formula: C₇H₁₂N₂O₂
MW: 156.19 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03617955
CAS Number: 258281-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR