Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate 95%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Ethylpyrazolo[1,5-a]pyridine-3-carboxylate

OR14354-250MG OR14354-1G
76567-002EA 52 USD
76567-002 76567-000
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate 95%
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190.1986 g/mol
Boiling Pt: 134…135 °C/2.3mmHg
Density: 1.23 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06411542
CAS Number: 16205-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR