5-Nitro-2-phenylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR14228-1G
76566-864EA 189 USD
76566-864
5-Nitro-2-phenylpyridine
5-Nitro-2-phenylpyridine
Formula: C₁₁H₈N₂O₂
Melting Pt: 116…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114193
CAS Number: 89076-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR