5-Amino-2-(4-methoxyphenyl)pyridine

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 6-(4-Methoxyphenyl)pyridin-3-amine

OR14219-250MG
76566-856EA 47 USD
76566-856
5-Amino-2-(4-methoxyphenyl)pyridine
5-Amino-2-(4-methoxyphenyl)pyridine
Formula: C₁₂H₁₂N₂O
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD05864840
CAS Number: 52057-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR