4-Isopropoxyphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Isopropoxybenzeneboronic acid

OR14157-5G OR14157-25G OR14157-100G
76566-788EA 37 USD
76566-788 76566-790 76566-786
4-Isopropoxyphenylboronic acid
4-Isopropoxyphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
Melting Pt: 150…154 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03427051
CAS Number: 153624-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR