Methyl-3-(methylamino)-4-nitrobenzenecarboxylate

Warning

Synonyms: Methyl 3-(methylamino)-4-nitrobenzoate

OR14145-1G OR14145-5G
76563-296EA 190.55 USD
76563-296 76562-968
Methyl-3-(methylamino)-4-nitrobenzenecarboxylate
Methyl-3-(methylamino)-4-nitrobenzenecarboxylate
Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
Melting Pt: 135…136 °C
Storage Temperature: Ambient
MDL Number: MFCD08689705
CAS Number: 251643-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR