Methyl-3-aminoisonicotinate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 3-aminopyridine-4-carboxylateMethyl 3-aminoisonicotinate

OR14139-25G OR14139-100G OR14139-5G
76566-756EA 342 USD
76566-756 76566-752 76566-754
Methyl-3-aminoisonicotinate
Methyl 3-aminoisonicotinate
Formula: C₇H₈N₂O₂
MW: 152.20 g/mol
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD02082535
CAS Number: 55279-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR