2,2-Dimethoxy-1-piperidino-1-ethanone

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2,2-Dimethoxy-1-(piperidin-1-yl)ethanone

OR14116-250MG OR14116-1G
76566-722EA 37 USD
76566-722 76566-724
2,2-Dimethoxy-1-piperidino-1-ethanone
2,2-Dimethoxy-1-piperidino-1-ethanone
Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
Boiling Pt: 72 °C/0.01mmHg
Storage Temperature: Ambient
MDL Number: MFCD08689667
CAS Number: 16695-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR