5-Bromo-6-phenyl-3(2H)-pyridazinone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromo-6-phenylpyridazin-3(2H)-one

OR14091-10G OR14091-1G OR14091-5G
76566-706EA 16 USD
76566-706 76566-708 76566-710
5-Bromo-6-phenyl-3(2H)-pyridazinone
5-Bromo-6-phenyl-3(2H)-pyridazinone
Formula: C₁₀H₇BrN₂O
MW: 251.09 g/mol
Melting Pt: 234…237 °C
Storage Temperature: Ambient
MDL Number: MFCD02083298
CAS Number: 90766-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR